This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBALD2 SIRNA Assays ›
Gene Symbol:
UBALD2
Gene Name:
UBA like domain containing 2
Gene Aliases:
FAM100B
Chromosome Location:
Chr.17: 76265205 - 76271298 on Build GRCh38
UniGene ID:
Hs.744981
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49433

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBALD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182565.3 NP_872371.1 3 911
GenBank mRNA BC035511.2 - 3 749 5091979
BC060859.1 - 3 914 30333976
CT004974.1 - 3 748

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0