This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM177A1 SIRNA Assays ›
Gene Symbol:
FAM177A1
Gene Name:
family with sequence similarity 177 member A1
Gene Aliases:
C14orf24
Chromosome Location:
Chr.14: 35044907 - 35083383 on Build GRCh38
UniGene ID:
Hs.446357
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49321

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM177A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079519.1 NP_001072987.1 4, 5 722
NM_001289022.1 NP_001275951.1 3, 4 445
NM_173607.3 NP_775878.2 2, 3 392
GenBank mRNA AK096173.1 - 3, 4 333
AL833508.1 - 2, 3 306
BC029559.1 - 3, 4 314 5310901
BG700755.1 - 3, 4 445
BX640884.1 - 2, 3 395
BX640984.1 - 2, 3 343

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0