This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C12ORF40 SIRNA Assays ›
Gene Symbol:
C12orf40
Gene Name:
chromosome 12 open reading frame 40
Gene Aliases:
HEL-206, HEL-S-94
Chromosome Location:
Chr.12: 39626167 - 39908300 on Build GRCh38
UniGene ID:
Hs.648205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49250

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C12orf40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031748.3 NP_001026918.2 6 649
NM_001319247.1 NP_001306176.1 6 649
NR_135051.1 5 510
XM_005268806.3 XP_005268863.1 5 459
XM_011538231.2 XP_011536533.1 5 686
GenBank mRNA AK097445.1 - 5 459
AL705293.1 - 6 625
BC038754.1 - 6 646 5269851
BC048120.1 - 6 633 5266211
BI461967.1 - 6 650
EU668334.1 - 5 459
GQ472216.1 - 5 459

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0