This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLEC9A SIRNA Assays ›
Gene Symbol:
CLEC9A
Gene Name:
C-type lectin domain family 9 member A
Gene Aliases:
CD370, DNGR-1, DNGR1, UNQ9341
Chromosome Location:
Chr.12: 10030677 - 10066030 on Build GRCh38
UniGene ID:
Hs.531189
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49230

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CLEC9A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207345.3 NP_997228.1 4, 5 712
GenBank mRNA AY358265.1 - 4, 5 712
EU339276.1 - 1, 2 99

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
positive regulation of cytokine secretion
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm