This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNHD1 SIRNA Assays ›
Gene Symbol:
DNHD1
Gene Name:
dynein heavy chain domain 1
Gene Aliases:
C11orf47, CCDC35, DHCD1, DNHD1L
Chromosome Location:
Chr.11: 6497296 - 6572025 on Build GRCh38
UniGene ID:
Hs.720080
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49176

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DNHD1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144666.2 NP_653267.2 4 1431
NM_173589.3 NP_775860.3 3 1534
GenBank mRNA AK096944.1 - 4 515
BC111765.1 - 2 1284
BX648742.1 - 3 1529

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
microtubule motor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0