This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF169 SIRNA Assays ›
Gene Symbol:
RNF169
Gene Name:
ring finger protein 169
Gene Aliases:
-
Chromosome Location:
Chr.11: 74748856 - 74842413 on Build GRCh38
UniGene ID:
Hs.370145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s48510

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF169
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098638.1 NP_001092108.1 6 1514
XM_011544889.2 XP_011543191.1 6 1589
GenBank mRNA AB082522.1 - 6 1514
BX640750.1 - 1 418

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
protein ubiquitination
negative regulation of double-strand break repair
zinc ion binding
ligase activity
nucleosome binding
K63-linked polyubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0