This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIF6 SIRNA Assays ›
Gene Symbol:
KIF6
Gene Name:
kinesin family member 6
Gene Aliases:
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location:
Chr.6: 39335100 - 39725405 on Build GRCh38
UniGene ID:
Hs.588202
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s48052

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIF6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289020.1 NP_001275949.1 8, 9 1084
NM_001289021.1 NP_001275950.1 8, 9 1084
NM_145027.4 NP_659464.3 8, 9 1084
XM_005248904.4 XP_005248961.1 8, 9 1084
XM_011514357.2 XP_011512659.1 8, 9 1009
XM_011514358.2 XP_011512660.1 8, 9 1009
XM_011514359.2 XP_011512661.1 8, 9 1009
XM_011514361.2 XP_011512663.1 8, 9 1084
XM_017010427.1 XP_016865916.1 7, 8 974
XM_017010428.1 XP_016865917.1 3, 4 434
GenBank mRNA AK131471.1 - 8, 9 1084
BC117307.1 - 8, 9 1014 40125858
BC143444.1 - 8, 9 1014 9051952
BC143445.1 - 8, 9 1014 9051953
BX649045.1 - 5, 6 664

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATPase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0