This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC73 SIRNA Assays ›
Gene Symbol:
LRRC73
Gene Name:
leucine rich repeat containing 73
Gene Aliases:
C6orf154
Chromosome Location:
Chr.6: 43506965 - 43510343 on Build GRCh38
UniGene ID:
Hs.445552
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s48047

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC73
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012974.2 NP_001012992.1 5 1282
NM_001271882.1 NP_001258811.1 5 590
GenBank mRNA AK126786.1 - 4 502
BC107130.2 - 5 490
BC108661.1 - 5 1282 5240367

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024360, XM_008766878, BC128712, AB210855.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0