This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATS1 SIRNA Assays ›
Gene Symbol:
SPATS1
Gene Name:
spermatogenesis associated serine rich 1
Gene Aliases:
DDIP, SPATA8, SRSP1
Chromosome Location:
Chr.6: 44342660 - 44377167 on Build GRCh38
UniGene ID:
Hs.135283
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s48038

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPATS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145026.3 NP_659463.1 9 1013
GenBank mRNA AK058171.1 - 8 1259
BC100956.2 - 8 1068 40004668
BC100957.2 - 8 1068 40004669
BC100958.2 - 8 1068 40004672
BC100959.2 - 9 1162 40004675
BC110065.1 - 8 1068 40004676
BC111414.1 - 8 1068 40004677

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0