This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STRA13 SIRNA Assays ›
Gene Symbol:
STRA13
Gene Name:
stimulated by retinoic acid 13
Gene Aliases:
CENP-X, CENPX, D9, FAAP10, MHF2
Chromosome Location:
Chr.17: 82018703 - 82022930 on Build GRCh38
UniGene ID:
Hs.37616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s47346

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STRA13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271006.1 NP_001257935.1 5 408
NM_001271007.1 NP_001257936.1 4 339
NM_144998.3 NP_659435.2 4 354
XM_005256339.2 XP_005256396.1 5 560
XM_017024326.1 XP_016879815.1 3 761
XM_017024327.1 XP_016879816.1 4 633
XM_017024328.1 XP_016879817.1 4 502
XM_017024329.1 XP_016879818.1 3 448
GenBank mRNA BC009571.1 - 4 323 4333630
BC011610.1 - 4 290
BI837753.1 - 4 322
BQ648002.1 - 1 43
CB110145.1 - 5 417
HY110543.1 - 5 398
U95006.1 - 4 308
U95007.1 - 4 293

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

resolution of meiotic recombination intermediates
mitotic nuclear division
replication fork processing
positive regulation of protein ubiquitination
CENP-A containing nucleosome assembly
interstrand cross-link repair
cell division
kinetochore assembly
DNA binding
double-stranded DNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0