This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF92 SIRNA Assays ›
Gene Symbol:
ZNF92
Gene Name:
zinc finger protein 92
Gene Aliases:
HEL-203, HPF12, HTF12, TF12
Chromosome Location:
Chr.7: 65373799 - 65401135 on Build GRCh38
UniGene ID:
Hs.9521
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46693

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF92
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287532.1 NP_001274461.1 3 996
NM_001287533.1 NP_001274462.1 5 1214
NM_001287534.1 NP_001274463.1 2 869
NM_007139.3 NP_009070.2 3 965
NM_152626.3 NP_689839.1 4 1092
GenBank mRNA AK091618.1 - 4 1010
BC036439.1 - 3 888 4817733
BC040594.1 - 4 1034 5269950
BX647875.1 - 2 814
EU668348.1 - 4 1036

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0