This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FNDC7 SIRNA Assays ›
Gene Symbol:
FNDC7
Gene Name:
fibronectin type III domain containing 7
Gene Aliases:
-
Chromosome Location:
Chr.1: 108712934 - 108742749 on Build GRCh38
UniGene ID:
Hs.258253
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46456

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNDC7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144937.1 NP_001138409.1 7 1216
GenBank mRNA AK093157.1 - 3 542
BC057762.1 - 3 589
BC130421.1 - 3 542 40146452

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header