This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC39B SIRNA Assays ›
Gene Symbol:
TTC39B
Gene Name:
tetratricopeptide repeat domain 39B
Gene Aliases:
C9orf52
Chromosome Location:
Chr.9: 15167204 - 15307360 on Build GRCh38
UniGene ID:
Hs.563630
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46040

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTC39B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168339.1 NP_001161811.1 14 1383
NM_001168340.1 NP_001161812.1 13 1350
NM_001168341.1 NP_001161813.1 12 1182
NM_001168342.1 NP_001161814.1 12 1264
NM_152574.2 NP_689787.2 14 1389
XM_011517732.2 XP_011516034.1 13 1186
XM_017014310.1 XP_016869799.1 16 1752
XM_017014311.1 XP_016869800.1 15 1684
XM_017014312.1 XP_016869801.1 13 1254
GenBank mRNA AK091187.1 - 14 1275
AK093459.1 - 14 1456
AK298581.1 - 12 1037
AK298998.1 - 13 1322
AK301427.1 - 12 1180
AK310482.1 - 14 1389
BC038592.1 - 1 99
BC042417.1 - 13 1256
BC142985.1 - 14 1362 8860428

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header