This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM160A1 SIRNA Assays ›
Gene Symbol:
FAM160A1
Gene Name:
family with sequence similarity 160 member A1
Gene Aliases:
-
Chromosome Location:
Chr.4: 151409143 - 151670503 on Build GRCh38
UniGene ID:
Hs.633810
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s458550

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM160A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109977.1 NP_001103447.1 14 3797
XM_005263199.1 XP_005263256.1 13 3672
XM_011532220.1 XP_011530522.1 12 3550
XM_011532221.2 XP_011530523.1 14 3870
XM_011532222.2 XP_011530524.1 13 3792
XM_011532223.2 XP_011530525.1 12 3667
XM_011532224.2 XP_011530526.1 12 3366
XM_017008583.1 XP_016864072.1 6 2127
GenBank mRNA AK125363.1 - 10 2031

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0