This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC112 SIRNA Assays ›
Gene Symbol:
CCDC112
Gene Name:
coiled-coil domain containing 112
Gene Aliases:
MBC1
Chromosome Location:
Chr.5: 115267188 - 115296761 on Build GRCh38
UniGene ID:
Hs.436121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45815

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC112
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040440.2 NP_001035530.1 3 493
NM_152549.2 NP_689762.2 3 539
XM_005271908.1 XP_005271965.1 3 487
GenBank mRNA AJ580807.1 - 3 392
BC031242.1 - 3 517 5265792

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109124.
This siRNA also targets the following transcripts in Mouse: NM_001160399, CJ174591, DT929545.

Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0