This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM167A SIRNA Assays ›
Gene Symbol:
TMEM167A
Gene Name:
transmembrane protein 167A
Gene Aliases:
TMEM167
Chromosome Location:
Chr.5: 83052846 - 83077453 on Build GRCh38
UniGene ID:
Hs.355606
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45774

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM167A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174909.4 NP_777569.1 4 297
GenBank mRNA BC026285.1 - 4 187 4762531
BC028585.1 - 4 221 4836971
BC070122.1 - 4 215
BC107575.1 - 4 208 4282548
BX647979.1 - 4 197
DA100343.1 - 4 340

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm