This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRMT44 SIRNA Assays ›
Gene Symbol:
TRMT44
Gene Name:
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases:
C4orf23, METTL19, TRM44
Chromosome Location:
Chr.4: 8440774 - 8512531 on Build GRCh38
UniGene ID:
Hs.566191
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45753

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRMT44
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152544.2 NP_689757.2 8 1474
XM_005247950.3 XP_005248007.1 8 1471
XM_011513406.2 XP_011511708.1 8 1503
XM_011513407.2 XP_011511709.1 8 1503
XM_011513408.1 XP_011511710.1 9 1615
XM_011513409.2 XP_011511711.1 9 1586
GenBank mRNA AK093044.1 - 4 480
BC035655.1 - 8 974 5588410

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

tRNA methylation
tRNA (uracil) methyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0