This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPM1K SIRNA Assays ›
Gene Symbol:
PPM1K
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1K
Gene Aliases:
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07
Chromosome Location:
Chr.4: 88257609 - 88284831 on Build GRCh38
UniGene ID:
Hs.43744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45746

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPM1K
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152542.4 NP_689755.3 6 1389
XM_006714111.3 XP_006714174.1 6 1450
XM_017007803.1 XP_016863292.1 6 1037
XM_017007804.1 XP_016863293.1 7 1577
XM_017007805.1 XP_016863294.1 8 1577
XM_017007806.1 XP_016863295.1 6 910
XM_017007807.1 XP_016863296.1 5 628
GenBank mRNA AK054678.1 - 6 1117
AK314417.1 - 6 1114
AL834271.1 - 6 1108
AX766363.1 - 5 904
AY157615.1 - 6 1117
AY435431.1 - 6 1294
AY994097.1 - 6 1125
BC037552.1 - 6 1153 5227317

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
branched-chain amino acid catabolic process
protein serine/threonine phosphatase activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0