This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CMTM8 SIRNA Assays ›
Gene Symbol:
CMTM8
Gene Name:
CKLF like MARVEL transmembrane domain containing 8
Gene Aliases:
CKLFSF8, CKLFSF8-V2
Chromosome Location:
Chr.3: 32238679 - 32370325 on Build GRCh38
UniGene ID:
Hs.154986
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45676

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CMTM8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320308.1 NP_001307237.1 3 590
NM_178868.4 NP_849199.2 4 764
XM_011533416.2 XP_011531718.1 5 564
XM_017005779.1 XP_016861268.1 4 1084
XM_017005780.1 XP_016861269.1 4 438
GenBank mRNA AF474370.2 - 4 764
AY598783.1 - 3 590
BC041390.2 - 4 719 5276695
CB070190.1 - 2 155

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
cytokine activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0