This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other XXYLT1 SIRNA Assays ›
Gene Symbol:
XXYLT1
Gene Name:
xyloside xylosyltransferase 1
Gene Aliases:
C3orf21
Chromosome Location:
Chr.3: 195068276 - 195271166 on Build GRCh38
UniGene ID:
Hs.478741
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45639

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: XXYLT1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308069.1 NP_001294998.1 4 1217
NM_152531.4 NP_689744.3 4 1680
XM_005269286.4 XP_005269343.1 4 4065
XM_011512448.2 XP_011510750.1 5 1492
XM_017005750.1 XP_016861239.1 3 4453
GenBank mRNA AK057046.1 - 5 1425
AK075551.1 - 4 1648
AK092474.1 - 2 957
BC019036.2 - 4 1589 4591786
BC039067.1 - 4 1662 5185557

Back To Top

More Information


Gene Ontology Categories:

Process(es)

transferase activity, transferring pentosyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0