This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALS2CR11 SIRNA Assays ›
Gene Symbol:
ALS2CR11
Gene Name:
amyotrophic lateral sclerosis 2 chromosome region candidate 11
Gene Aliases:
-
Chromosome Location:
Chr.2: 201487421 - 201619182 on Build GRCh38
UniGene ID:
Hs.335788
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45547

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ALS2CR11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168216.1 NP_001161688.1 13 1447
NM_001168221.1 NP_001161693.1 16 5382
NM_152525.5 NP_689738.3 15 1791
XM_006712331.3 XP_006712394.1 15 1794
XM_006712332.3 XP_006712395.1 14 1719
GenBank mRNA AB053313.1 - 2 1139
AK058080.1 - 13 1419
AK097651.1 - 2 2397
BC030659.2 - 15 1784 4838801
BX648732.1 - 16 5375

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0