This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2U SIRNA Assays ›
Gene Symbol:
UBE2U
Gene Name:
ubiquitin conjugating enzyme E2 U (putative)
Gene Aliases:
-
Chromosome Location:
Chr.1: 64202802 - 64267381 on Build GRCh38
UniGene ID:
Hs.388746
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45194

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBE2U
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152489.1 NP_689702.1 8 867
XM_005270498.4 XP_005270555.1 8 1661
XM_006710379.3 XP_006710442.1 8 802
XM_011540764.2 XP_011539066.1 9 1602
XM_017000379.1 XP_016855868.1 6 429
XM_017000380.1 XP_016855869.1 7 473
XM_017000381.1 XP_016855870.1 6 382
XM_017000382.1 XP_016855871.1 5 295
XM_017000383.1 XP_016855872.1 6 388
XM_017000384.1 XP_016855873.1 5 286
XM_017000385.1 XP_016855874.1 4 255
GenBank mRNA BC029895.1 - 8 867 5168406

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
histone ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0