This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATA8 SIRNA Assays ›
Gene Symbol:
SPATA8
Gene Name:
spermatogenesis associated 8
Gene Aliases:
SRG8
Chromosome Location:
Chr.15: 96783407 - 96785615 on Build GRCh38
UniGene ID:
Hs.326528
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44804

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPATA8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304804.1 NP_001291733.1 2 135
NM_001304805.1 NP_001291734.1 3 284
NM_001304806.1 NP_001291735.1 2 198
NM_173499.4 NP_775770.1 2 353
XM_011521265.2 XP_011519567.1 2 400
GenBank mRNA AY489187.1 - 2 317
BC033979.2 - 2 290 5295377
BC066292.1 - 2 312 5271997
BC112390.1 - 1 681 5168326
HY024478.1 - 2 161
HY034703.1 - 3 281
HY047940.1 - 2 135

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0