This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OR10A5 SIRNA Assays ›
Gene Symbol:
OR10A5
Gene Name:
olfactory receptor family 10 subfamily A member 5
Gene Aliases:
JCG6, OR10A1, OR11-403
Chromosome Location:
Chr.11: 6845683 - 6846636 on Build GRCh38
UniGene ID:
Hs.447478
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44602

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR10A5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178168.1 NP_835462.1 1 820
GenBank mRNA AF324499.1 - 1 820
BC093651.1 - 1 851 7939496
BC093653.1 - 1 851 7939498

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0