This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACSM6 SIRNA Assays ›
Gene Symbol:
ACSM6
Gene Name:
acyl-CoA synthetase medium-chain family member 6
Gene Aliases:
C10orf129, bA310E22.3
Chromosome Location:
Chr.10: 95194200 - 95228929 on Build GRCh38
UniGene ID:
Hs.134229
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44494

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ACSM6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207321.2 NP_997204.2 6 944
GenBank mRNA AK126134.1 - 6 934
BC063654.1 - 3 333
BC104210.1 - 5 828
BC104211.2 - 5 828
BC112988.1 - 6 909
BC148184.1 - 5 828

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
regulation of protein stability
positive regulation of protein targeting to mitochondrion
ATP binding
GTP binding
metal ion binding
butyrate-CoA ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0