This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TLDC2 SIRNA Assays ›
Gene Symbol:
TLDC2
Gene Name:
TBC/LysM-associated domain containing 2
Gene Aliases:
C20orf118
Chromosome Location:
Chr.20: 36873071 - 36894235 on Build GRCh38
UniGene ID:
Hs.472630
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44369

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TLDC2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304783.1 NP_001291712.1 4 428
NM_080628.2 NP_542195.1 5 524
XM_017027672.1 XP_016883161.1 5 514
XM_017027673.1 XP_016883162.1 5 543
XM_017027674.1 XP_016883163.1 4 364
GenBank mRNA AK123544.1 - 5 478
BC127688.1 - 5 524 40127440
BC130646.1 - 5 513 40146677
BC130648.1 - 5 513 40146679
BG163960.1 - 23
BY994247.1 - 4 416

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0