This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASB9 SIRNA Assays ›
Gene Symbol:
ASB9
Gene Name:
ankyrin repeat and SOCS box containing 9
Gene Aliases:
-
Chromosome Location:
Chr.X: 15243987 - 15270476 on Build GRCh38
UniGene ID:
Hs.19404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44287

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASB9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031739.2 NP_001026909.1 3, 4 587
NM_001168530.1 NP_001162002.1 3, 4 587
NM_001168531.1 NP_001162003.1 4, 5 774
NM_024087.2 NP_076992.1 3, 4 587
XM_005274446.1 XP_005274503.1 4, 5 774
XM_017029283.1 XP_016884772.1 3, 4 554
XM_017029285.1 XP_016884774.1 3, 4 554
XM_017029286.1 XP_016884775.1 3, 4 554
GenBank mRNA AK000643.1 - 3, 4 477
AK292270.1 - 4, 5 774
AL080091.2 - 3, 4 303
BC001244.1 - 3, 4 467 3462727
BC013172.1 - 3, 4 537 3917449

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
intracellular signal transduction
positive regulation of protein catabolic process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0