This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASB9 SIRNA Assays ›
Gene Symbol:
ASB9
Gene Name:
ankyrin repeat and SOCS box containing 9
Gene Aliases:
-
Chromosome Location:
Chr.X: 15243987 - 15270476 on Build GRCh38
UniGene ID:
Hs.19404
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44285

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASB9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031739.2 NP_001026909.1 7 1094
NM_001168530.1 NP_001162002.1 7 1125
NM_001168531.1 NP_001162003.1 8 1342
NM_024087.2 NP_076992.1 7 1155
XM_005274446.1 XP_005274503.1 8 1281
XM_011545458.1 XP_011543760.1 8 1194
XM_011545460.1 XP_011543762.1 8 1255
XM_017029283.1 XP_016884772.1 7 1061
XM_017029284.1 XP_016884773.1 7 974
XM_017029285.1 XP_016884774.1 7 1122
XM_017029286.1 XP_016884775.1 7 1126
GenBank mRNA AK000643.1 - 7 1045
AK292270.1 - 8 1342
AL080091.2 - 7 871
BC001244.1 - 7 1005 3462727
BC013172.1 - 7 1044 3917449

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
intracellular signal transduction
positive regulation of protein catabolic process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header