This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MUC17 SIRNA Assays ›
Gene Symbol:
MUC17
Gene Name:
mucin 17, cell surface associated
Gene Aliases:
MUC-17, MUC-3, MUC3
Chromosome Location:
Chr.7: 101020083 - 101058859 on Build GRCh38
UniGene ID:
Hs.271819
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44269

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MUC17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040105.1 NP_001035194.1 3 675
NR_133665.1 3 675
GenBank mRNA AJ606307.1 - 3 675
AJ606308.1 - 3 675

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

O-glycan processing
cellular homeostasis
protein binding
PDZ domain binding
extracellular matrix constituent, lubricant activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0