This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RALYL SIRNA Assays ›
Gene Symbol:
RALYL
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
HNRPCL3
Chromosome Location:
Chr.8: 84182629 - 84921844 on Build GRCh38
UniGene ID:
Hs.121663
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44079

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RALYL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100391.1 NP_001093861.1 4, 5 546
NM_001100392.1 NP_001093862.1 5, 6 789
NM_001100393.1 NP_001093863.1 4, 5 757
NM_001287244.1 NP_001274173.1 3, 4 251
NM_173848.5 NP_776247.3 4, 5 1470
XM_017013059.1 XP_016868548.1 6, 7 840
XM_017013060.1 XP_016868549.1 4, 5 2990
XM_017013061.1 XP_016868550.1 4, 5 2142
XM_017013062.1 XP_016868551.1 6, 7 949
XM_017013063.1 XP_016868552.1 4, 5 712
XM_017013064.1 XP_016868553.1 5, 6 1063
XM_017013065.1 XP_016868554.1 4, 5 611
XM_017013072.1 XP_016868561.1 4, 5 698
XM_017013073.1 XP_016868562.1 4, 5 698
XM_017013074.1 XP_016868563.1 4, 5 611
XM_017013076.1 XP_016868565.1 4, 5 491
XM_017013077.1 XP_016868566.1 5, 6 1741
XM_017013078.1 XP_016868567.1 4, 5 757
XM_017013079.1 XP_016868568.1 5, 6 740
XM_017013083.1 XP_016868572.1 3, 4 412
XM_017013085.1 XP_016868574.1 4, 5 294
XM_017013086.1 XP_016868575.1 3, 4 251
XM_017013087.1 XP_016868576.1 3, 4 251
GenBank mRNA AF495529.1 - 4, 5 1470
AF495530.1 - 4, 5 687
AK094543.1 - 4, 5 701
AK095560.1 - 5, 6 740
AK123437.1 - 3, 4 251
BC031090.1 - 4, 5 535 5311935
DN989439.1 - 4, 5 551

Back To Top

More Information


Gene Ontology Categories:

Process(es)

nucleotide binding
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0