This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C5ORF47 SIRNA Assays ›
Gene Symbol:
C5orf47
Gene Name:
chromosome 5 open reading frame 47
Gene Aliases:
-
Chromosome Location:
Chr.5: 173989159 - 174007524 on Build GRCh38
UniGene ID:
Hs.131469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s43782

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C5orf47
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144954.1 NP_001138426.1 5 827
XM_011534431.1 XP_011532733.1 4 721
GenBank mRNA BC034971.1 - 5 793
BC047225.1 - 5 782
BC053953.1 - 5 782
BC057842.1 - 5 768
BC066964.1 - 5 789
BC092489.1 - 5 816
BC121167.1 - 4 448
BC121168.1 - 4 448

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0