This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM131A SIRNA Assays ›
Gene Symbol:
FAM131A
Gene Name:
family with sequence similarity 131 member A
Gene Aliases:
C3orf40, FLAT715, PRO1378
Chromosome Location:
Chr.3: 184335924 - 184346275 on Build GRCh38
UniGene ID:
Hs.591307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s43603

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM131A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171093.1 NP_001164564.1 6 934
NM_144635.4 NP_653236.3 6 1052
XM_005247113.2 XP_005247170.1 7 1082
XM_005247114.3 XP_005247171.1 5 878
XM_011512413.2 XP_011510715.1 5 1120
GenBank mRNA AY358435.1 - 3 2180
BC026221.1 - 5 691 4399325
BC037299.1 - 6 931 4825140

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0