This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4348
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
3
395
NM_001256304.1
NP_001243233.1
2
132
NM_001256308.2
NP_001243237.1
2
266
NM_001320932.1
NP_001307861.1
3
395
NM_001320933.1
NP_001307862.1
3
395
NM_001320934.1
NP_001307863.1
3
395
NM_001320935.1
NP_001307864.1
3
395
NM_001320936.1
NP_001307865.1
3
395
NM_001320937.1
NP_001307866.1
3
395
NM_021907.4
NP_068707.1
3
395
NM_033147.3
NP_149159.2
3
395
NM_033148.3
NP_149160.1
3
395
NM_183360.2
NP_899204.1
3
395
NM_183361.2
NP_899205.1
3
395
XM_005264182.1
XP_005264239.1
3
312
XM_011532671.1
XP_011530973.1
3
312
XM_011532672.2
XP_011530974.1
3
312
XM_011532673.1
XP_011530975.1
3
312
XM_011532674.1
XP_011530976.1
3
312
XM_011532675.1
XP_011530977.1
3
312
XM_011532676.1
XP_011530978.1
3
312
XM_011532677.2
XP_011530979.1
2
768
XM_011532679.1
XP_011530981.1
3
312
XM_011532680.2
XP_011530982.1
3
319
XM_011532681.2
XP_011530983.1
3
312
XM_017003529.1
XP_016859018.1
3
312
XM_017003530.1
XP_016859019.1
3
312
XM_017003531.1
XP_016859020.1
3
312
XM_017003532.1
XP_016859021.1
3
312
XM_017003533.1
XP_016859022.1
3
312
XM_017003534.1
XP_016859023.1
3
312
XM_017003535.1
XP_016859024.1
3
312
XM_017003536.1
XP_016859025.1
3
312
XM_017003538.1
XP_016859027.1
3
244
XM_017003539.1
XP_016859028.1
3
255
XM_017003540.1
XP_016859029.1
3
312
XM_017003541.1
XP_016859030.1
3
296
XM_017003542.1
XP_016859031.1
3
319
XM_017003543.1
XP_016859032.1
3
299
XM_017003544.1
XP_016859033.1
3
319
XM_017003545.1
XP_016859034.1
3
299
GenBank mRNA
AF022728.1
-
3
293
AK094494.1
-
3
258
AK225724.1
-
3
298
AK294174.1
-
3
300
AK294357.1
-
3
162
AK300019.1
-
3
272
AK301386.1
-
2
268
AL555818.3
-
3
309
BC016655.1
-
3
288
3868207
BC049366.1
-
3
386
4817552
BT009805.1
-
2
132
BU846095.1
-
1
73
DB460644.1
-
3
395
DQ160290.1
-
2
132
Y12712.1
-
3
315
Y15718.1
-
2
132
Y15722.1
-
2
132
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0