This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANO4 SIRNA Assays ›
Gene Symbol:
ANO4
Gene Name:
anoctamin 4
Gene Aliases:
TMEM16D
Chromosome Location:
Chr.12: 100717457 - 101128641 on Build GRCh38
UniGene ID:
Hs.58785
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42492

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANO4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286615.1 NP_001273544.1 24 2936
NM_001286616.1 NP_001273545.1 23 2504
NM_178826.3 NP_849148.2 23 2831
XM_011537911.2 XP_011536213.1 26 2931
XM_011537912.2 XP_011536214.1 26 2931
XM_011537913.2 XP_011536215.1 25 2886
XM_011537914.2 XP_011536216.1 23 2442
XM_011537915.2 XP_011536217.1 24 2754
XM_011537916.2 XP_011536218.1 24 2595
XM_017018808.1 XP_016874297.1 23 2442
XM_017018809.1 XP_016874298.1 25 3022
XM_017018810.1 XP_016874299.1 23 2684
XM_017018811.1 XP_016874300.1 22 2639
XM_017018812.1 XP_016874301.1 22 4657
XM_017018813.1 XP_016874302.1 21 2507
XM_017018814.1 XP_016874303.1 24 4896
XM_017018815.1 XP_016874304.1 24 4896
XM_017018816.1 XP_016874305.1 24 2979
XM_017018817.1 XP_016874306.1 24 4931
GenBank mRNA AK091540.1 - 14 1452
AK091591.1 - 23 2620
AK092596.1 - 11 1018
AK293810.1 - 24 2574
AK316105.1 - 24 2575
BC109308.2 - 23 2489 40035759

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chloride transport
lipid transport
ion transmembrane transport
calcium activated phosphatidylserine scrambling
calcium activated phosphatidylcholine scrambling
calcium activated galactosylceramide scrambling
chloride transmembrane transport
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0