This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF641 SIRNA Assays ›
Gene Symbol:
ZNF641
Gene Name:
zinc finger protein 641
Gene Aliases:
-
Chromosome Location:
Chr.12: 48334380 - 48351414 on Build GRCh38
UniGene ID:
Hs.23492
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42449

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF641
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172681.1 NP_001166152.1 5 565
NM_001172682.1 NP_001166153.1 4 471
NM_152320.2 NP_689533.2 6 738
XM_005268637.3 XP_005268694.1 4 518
XM_005268638.3 XP_005268695.1 5 758
XM_005268639.2 XP_005268696.1 4 409
XM_005268640.4 XP_005268697.1 5 738
XM_011537897.1 XP_011536199.1 5 738
XM_011537898.1 XP_011536200.1 5 666
XM_017018797.1 XP_016874286.1 4 562
XM_017018798.1 XP_016874287.1 5 592
XM_017018799.1 XP_016874288.1 4 472
GenBank mRNA AK055857.1 - 5 641
AK092740.1 - 5 1165
AK298064.1 - 4 463
AY842285.1 - 5 1192
BC018090.1 - 5 565 4791876

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0