This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DEPDC4 SIRNA Assays ›
Gene Symbol:
DEPDC4
Gene Name:
DEP domain containing 4
Gene Aliases:
DEP.4
Chromosome Location:
Chr.12: 100203669 - 100268094 on Build GRCh38
UniGene ID:
Hs.653118
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42411

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DEPDC4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319310.1 NP_001306239.1 3 687
NM_001319311.1 NP_001306240.1 3 687
NM_152317.3 NP_689530.1 3 687
NR_135073.1 3 687
XM_011537876.2 XP_011536178.1 2 290
XM_017018779.1 XP_016874268.1 3 687
XM_017018780.1 XP_016874269.1 3 687
XM_017018781.1 XP_016874270.1 3 687
XM_017018782.1 XP_016874271.1 3 580
XM_017018783.1 XP_016874272.1 3 815
XM_017018785.1 XP_016874274.1 3 687
GenBank mRNA AK090824.1 - 3 687
BC015117.1 - 3 587
BC100927.2 - 2 290
BC100928.2 - 3 486
BC100929.1 - 2 290
BC100930.1 - 3 687

Back To Top

More Information


Gene Ontology Categories:

Function(s)

intracellular signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header