This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM64EP SIRNA Assays ›
Gene Symbol:
TRIM64EP TRIM64 TRIM64DP
Gene Name:
tripartite motif containing 64E, pseudogene
tripartite motif containing 64
tripartite motif containing 64D, pseudogene
Gene Aliases:
-
C11orf28, TRIM64A
-
Chromosome Location:
Chr.11: 90057419 - 90062997 on Build GRCh38
Chromosome Location:
Chr.11: 89966068 - 89974147 on Build GRCh38
Chromosome Location:
Chr.11: 89776981 - 89782560 on Build GRCh38
UniGene ID:
Hs.445846
Hs.454490
Hs.742589
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s42356

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIM64EP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA JF968451.1 - 1 198
Gene Symbol: TRIM64
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136486.1 NP_001129958.1 1 205
XM_011542585.2 XP_011540887.1 2 294
Gene Symbol: TRIM64DP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA JF968450.1 - 1 198

Back To Top

More Information


Gene Ontology Categories:

Process(es)

zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0