This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CATSPER1 SIRNA Assays ›
Gene Symbol:
CATSPER1
Gene Name:
cation channel sperm associated 1
Gene Aliases:
CATSPER, SPGF7
Chromosome Location:
Chr.11: 66016752 - 66026518 on Build GRCh38
UniGene ID:
Hs.189105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42102

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CATSPER1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053054.3 NP_444282.3 1 1341
GenBank mRNA AF407333.1 - 1 1203
BC032950.2 - 1 1340 4822261
BC036522.2 - 1 1303 5268567

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
fusion of sperm to egg plasma membrane
cell differentiation
sperm motility
response to progesterone
regulation of ion transmembrane transport
sperm-egg recognition
regulation of calcium ion transport
regulation of cilium beat frequency involved in ciliary motility
calcium ion transmembrane transport
membrane depolarization during action potential
calcium activated cation channel activity
voltage-gated calcium channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0