This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLNK SIRNA Assays ›
Gene Symbol:
CLNK
Gene Name:
cytokine dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.4: 10489899 - 10734853 on Build GRCh38
UniGene ID:
Hs.678910
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42023

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CLNK
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052964.2 NP_443196.2 8, 9 591
XM_011513775.2 XP_011512077.1 8, 9 822
XM_017007684.1 XP_016863173.1 8, 9 822
XM_017007685.1 XP_016863174.1 8, 9 822
GenBank mRNA AB032369.1 - 6, 7 400
AB110420.1 - 8, 9 694
AK093001.1 - 8, 9 593
BC029887.1 - 8, 9 621
BC137553.1 - 8, 9 520 9021553
BC137556.1 - 8, 9 520 9021556

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of signal transduction
intracellular signal transduction
SH3/SH2 adaptor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0