This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPECC1 SIRNA Assays ›
Gene Symbol:
SPECC1
Gene Name:
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases:
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location:
Chr.17: 20009301 - 20319026 on Build GRCh38
UniGene ID:
Hs.431045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s40960

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPECC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033553.2 NP_001028725.1 4 616
NM_001033554.2 NP_001028726.1 2 525
NM_001033555.2 NP_001028727.1 2 525
NM_001243438.1 NP_001230367.1 2 525
NM_001243439.1 NP_001230368.1 4 675
NM_152904.4 NP_690868.3 4 616
XM_011524075.1 XP_011522377.1 3 568
XM_017025317.1 XP_016880806.1 3 568
XM_017025318.1 XP_016880807.1 3 568
XM_017025319.1 XP_016880808.1 4 710
XM_017025320.1 XP_016880809.1 4 710
XM_017025321.1 XP_016880810.1 2 525
XM_017025322.1 XP_016880811.1 3 568
XM_017025323.1 XP_016880812.1 3 568
XM_017025324.1 XP_016880813.1 3 568
XM_017025325.1 XP_016880814.1 4 710
GenBank mRNA AB041533.1 - 4 616
AK295093.1 - 4 648
AK295811.1 - 1 152
AK304186.1 - 2 447
AK314341.1 - 4 691
AY816326.1 - 2 430
AY816327.1 - 2 430
AY816328.1 - 4 616
AY816329.1 - 4 616
BC021123.1 - 2 423 3614358
BC050058.1 - 2 360 5277450
BP309862.1 - 1 178
DC336225.1 - 2 530

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039017, XM_006246503, XM_008767822, XM_008767823, XM_008767821, and others.
This siRNA also targets the following transcripts in Mouse: NM_001281818, NM_001029936, AK220465, AK143663, AK046469, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0