This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1ORF105 SIRNA Assays ›
Gene Symbol:
C1orf105
Gene Name:
chromosome 1 open reading frame 105
Gene Aliases:
-
Chromosome Location:
Chr.1: 172418560 - 172468829 on Build GRCh38
UniGene ID:
Hs.517991
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s40934

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1orf105
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300760.1 NP_001287689.1 2 358
NM_139240.3 NP_640333.3 4 457
XM_005245608.2 XP_005245665.1 4 321
XM_005245610.3 XP_005245667.1 3 238
XM_006711630.2 XP_006711693.1 2 144
XM_011510152.2 XP_011508454.1 6 2666
XM_011510153.2 XP_011508455.1 5 2629
XM_011510154.2 XP_011508456.1 3 2499
XM_011510155.2 XP_011508457.1 6 2666
XM_011510156.2 XP_011508458.1 2 2408
XM_017002808.1 XP_016858297.1 4 255
GenBank mRNA AL035295.1 - 4 457
BC038410.1 - 4 455 5169137

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0