This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM4 SIRNA Assays ›
Gene Symbol:
TRIM4
Gene Name:
tripartite motif containing 4
Gene Aliases:
RNF87
Chromosome Location:
Chr.7: 99890407 - 99919600 on Build GRCh38
UniGene ID:
Hs.50749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s40101

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIM4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033017.3 NP_148977.2 5, 6 1012
NM_033091.2 NP_149082.1 4, 5 934
GenBank mRNA AF220023.1 - 6, 7 949
AF220024.1 - 4, 5 871
AK092892.1 - 4, 5 896
AK293565.1 - 6, 7 788
BC011763.2 - 4, 5 827 3617674
BC025949.1 - 4, 5 827 4620469

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
innate immune response
protein trimerization
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header