This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CGNL1 SIRNA Assays ›
Gene Symbol:
CGNL1
Gene Name:
cingulin like 1
Gene Aliases:
JACOP, PCING
Chromosome Location:
Chr.15: 57376487 - 57550727 on Build GRCh38
UniGene ID:
Hs.148989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s39740

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CGNL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252335.1 NP_001239264.1 16 3541
NM_032866.4 NP_116255.2 15 3426
XM_005254726.3 XP_005254783.1 15 3444
XM_005254727.4 XP_005254784.1 16 3822
XM_011522120.1 XP_011520422.1 16 3480
XM_011522121.1 XP_011520423.1 15 3603
XM_017022685.1 XP_016878174.1 15 4061
XM_017022686.1 XP_016878175.1 15 4059
GenBank mRNA AB051536.2 - 14 2585
AK027863.1 - 10 1336
AY274808.1 - 15 3424
BC118918.1 - 14 3367 40074275

Back To Top

More Information


Gene Ontology Categories:

Process(es)

motor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0