This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C9ORF3 SIRNA Assays ›
Gene Symbol:
C9orf3
Gene Name:
chromosome 9 open reading frame 3
Gene Aliases:
AOPEP, AP-O, APO, C90RF3, ONPEP
Chromosome Location:
Chr.9: 94726604 - 95148264 on Build GRCh38
UniGene ID:
Hs.434253
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s39631

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C9orf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193329.1 NP_001180258.1 7 1843
NM_032823.5 NP_116212.3 6 1629
XM_005252284.2 XP_005252341.1 6 1694
XM_006717306.1 XP_006717369.1 8 1991
XM_011519119.1 XP_011517421.1 8 1991
XM_011519120.2 XP_011517422.1 8 1991
XM_011519121.2 XP_011517423.1 8 1991
XM_011519122.2 XP_011517424.1 8 1991
XM_011519123.2 XP_011517425.1 8 1991
XM_011519127.1 XP_011517429.1 8 1991
XM_011519132.1 XP_011517434.1 5 717
XM_017015229.1 XP_016870718.1 8 2034
XM_017015230.1 XP_016870719.1 8 1991
XM_017015231.1 XP_016870720.1 6 1694
XM_017015232.1 XP_016870721.1 8 1991
XM_017015233.1 XP_016870722.1 8 2021
XM_017015234.1 XP_016870723.1 7 1824
XM_017015235.1 XP_016870724.1 7 1824
XM_017015236.1 XP_016870725.1 5 1527
XM_017015237.1 XP_016870726.1 5 1527
GenBank mRNA AJ560639.1 - 7 1708
AK027581.1 - 5 880
BC020194.1 - 7 1054 5020674
BC029616.1 - 6 1586 3877098
BX648970.1 - 6 929

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
peptide catabolic process
aminopeptidase activity
metallopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0