This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAXC SIRNA Assays ›
Gene Symbol:
FAXC
Gene Name:
failed axon connections homolog
Gene Aliases:
C6orf168, dJ273F20
Chromosome Location:
Chr.6: 99272915 - 99350081 on Build GRCh38
UniGene ID:
Hs.573245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s39154

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAXC
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032511.2 NP_115900.1 6 1461
XM_006715582.2 XP_006715645.1 4 661
XM_011536188.2 XP_011534490.1 6 978
GenBank mRNA AK057793.1 - 5 879
AK096480.1 - 6 1224
BC004869.2 - 6 1461 3834272
BC006515.2 - 3 392
BC011583.1 - 6 1461 4577456

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0