This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MFSD7 SIRNA Assays ›
Gene Symbol:
MFSD7
Gene Name:
major facilitator superfamily domain containing 7
Gene Aliases:
LP2561
Chromosome Location:
Chr.4: 676205 - 689468 on Build GRCh38
UniGene ID:
Hs.567612
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38613

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MFSD7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294341.1 NP_001281270.1 4 886
NM_032219.3 NP_115595.2 4 886
XM_005272299.4 XP_005272356.1 4 913
XM_006713918.1 XP_006713981.1 4 540
XM_011513568.1 XP_011511870.1 4 1690
XM_011513571.1 XP_011511873.1 5 710
XM_011513573.2 XP_011511875.1 4 913
XM_017008700.1 XP_016864189.1 4 913
GenBank mRNA AK292010.1 - 4 784
AY358401.1 - 4 588
BC030246.1 - 4 629 5216645

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transmembrane transport

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0