This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIAA1109 SIRNA Assays ›
Gene Symbol:
KIAA1109
Gene Name:
KIAA1109
Gene Aliases:
FSA, Tweek
Chromosome Location:
Chr.4: 122152333 - 122362759 on Build GRCh38
UniGene ID:
Hs.408142
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38584

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA1109
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015312.3 NP_056127.2 5 430
XM_005263282.1 XP_005263339.1 7 758
XM_005263287.1 XP_005263344.1 7 758
XM_006714344.1 XP_006714407.1 7 758
XM_011532319.1 XP_011530621.1 7 758
XM_011532320.2 XP_011530622.1 8 853
XM_011532321.2 XP_011530623.1 6 560
XM_011532322.1 XP_011530624.1 7 758
XM_011532323.1 XP_011530625.1 7 758
XM_011532324.1 XP_011530626.1 7 758
XM_011532325.1 XP_011530627.1 7 758
XM_011532326.1 XP_011530628.1 7 758
XM_011532327.1 XP_011530629.1 8 804
XM_017008695.1 XP_016864184.1 7 758
XM_017008696.1 XP_016864185.1 7 758
XM_017008697.1 XP_016864186.1 7 758
XM_017008699.1 XP_016864188.1 7 758
GenBank mRNA DQ335469.1 - 5 430

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
regulation of epithelial cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header