This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YPEL3 SIRNA Assays ›
Gene Symbol:
YPEL3
Gene Name:
yippee like 3
Gene Aliases:
-
Chromosome Location:
Chr.16: 30092314 - 30096216 on Build GRCh38
UniGene ID:
Hs.513491
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38147

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: YPEL3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145524.1 NP_001138996.1 2 240
NM_031477.4 NP_113665.3 1 886
GenBank mRNA AB098737.1 - 2 240
AF305622.1 - 2 243
AK022409.1 - 1 1109
AK026047.1 - 1 274
BC005009.1 - 2 196 3639517
BC050664.1 - 1 885 5748830

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cellular senescence
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0