This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO38 SIRNA Assays ›
Gene Symbol:
FBXO38
Gene Name:
F-box protein 38
Gene Aliases:
Fbx38, HMN2D, MOKA, SP329
Chromosome Location:
Chr.5: 148383935 - 148442836 on Build GRCh38
UniGene ID:
Hs.483772
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37602

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXO38
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271723.1 NP_001258652.1 11 1453
NM_030793.4 NP_110420.3 11 1453
XM_005268513.1 XP_005268570.1 11 1442
XM_006714797.1 XP_006714860.1 11 1442
XM_011537684.2 XP_011535986.1 11 1495
XM_017009899.1 XP_016865388.1 12 1590
XM_017009900.1 XP_016865389.1 12 1654
XM_017009901.1 XP_016865390.1 11 1507
XM_017009902.1 XP_016865391.1 11 1495
GenBank mRNA AF251055.1 - 4 391
AK024024.1 - 11 1374
AK308414.1 - 11 1402
BC005849.2 - 1 27
BC005873.2 - 1 27
BC033454.1 - 11 1386 4510274
BC050424.1 - 10 1358 6472693

Back To Top

More Information


Gene Ontology Categories:

Function(s)

positive regulation of neuron projection development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0