This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CTSG SIRNA Assays ›
Gene Symbol:
CTSG
Gene Name:
cathepsin G
Gene Aliases:
CATG, CG
Chromosome Location:
Chr.14: 24573518 - 24576260 on Build GRCh38
UniGene ID:
Hs.421724
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s3745

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CTSG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001911.2 NP_001902.1 4 395
XM_011536499.1 XP_011534801.1 4 415
GenBank mRNA AK225914.1 - 3 842
BC014460.1 - 4 395 4859911
CR456807.1 - 4 358
CR541704.1 - 4 358
M16117.1 - 4 366

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiotensin maturation
protein phosphorylation
proteolysis
immune response
protein processing
extracellular matrix disassembly
response to lipopolysaccharide
negative regulation of growth of symbiont in host
cellular protein metabolic process
positive regulation of immune response
defense response to fungus
neutrophil mediated killing of gram-positive bacterium
serine-type endopeptidase activity
protein binding
heparin binding
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0